- Efficient nucleophilic aromatic substitution between aryl nitrofluorides and alkynes.
Efficient nucleophilic aromatic substitution between aryl nitrofluorides and alkynes.
Organic letters (2007-06-08)
Patrick L DeRoy, Simon Surprenant, Megan Bertrand-Laperle, Christiane Yoakim
PMID17552534
RÉSUMÉ
The nucleophilic aromatic substitution reaction between electron-deficient aryl fluorides and terminal alkynes is shown to be efficiently promoted by sodium bis(trimethylsilyl)amide as a base. Moderate to excellent yields of 2-ethynylnitrobenzene products can be obtained under mild conditions.