Merck
Investigation on Furan Levels in Pressure-Cooked Foods.
International journal of food science (2013-01-01)
Adriana P Arisseto, Eduardo Vicente, Maria Cecília F Toledo
RÉSUMÉ
Furan is a food processing contaminant classified as possibly carcinogenic to humans. As the occurrence of furan has been reported in a variety of foods processed in sealed containers, the objective of this work was to investigate if the contaminant can be found in home-cooked foods prepared in a pressure cooker. For that, several foods including beans, soy beans, whole rice, beef, pork, potato, and cassava were pressure-cooked and analyzed for the furan content by gas chromatography coupled to mass spectrometry preceded by a headspace solid phase microextraction (HS-SPME-GC/MS). Furan was not found above the limit of quantification in the pressure-cooked samples. No furan has either been found in reheated samples after 24 hours under cold storage. Although levels up to 173 μg/kg were already reported for commercial canned/jarred foods, it seems that the cooking in a pressure cooker may not represent a concern in relation to the occurrence of furan in foods.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Furan-d4, ≥98 atom % D, ≥99% (CP), contains 0.025 wt. % BHT as stabilizer