Merck
NEDD9 and BCAR1 negatively regulate E-cadherin membrane localization, and promote E-cadherin degradation.
PloS one (2011-07-19)
Nadezhda Tikhmyanova, Erica A Golemis
RÉSUMÉ
The Cas scaffolding proteins (NEDD9/HEF1/CAS-L, BCAR1/p130Cas, EFSSIN, and HEPL/CASS4) regulate cell migration, division and survival, and are often deregulated in cancer. High BCAR1 expression is linked to poor prognosis in breast cancer patients, while upregulation of NEDD9 contributes to the metastatic behavior of melanoma and glioblastoma cells. Our recent work knocking out the single Drosophila Cas protein, Dcas, identified a genetic interaction with E-cadherin. As E-cadherin is often downregulated during epithelial-mesenchymal transition (EMT) prior to metastasis, if such an activity was conserved in mammals it might partially explain how Cas proteins promote aggressive tumor behavior. We here establish that Cas proteins negatively regulate E-cadherin expression in human mammary cells. Cas proteins do not affect E-cadherin transcription, but rather, BCAR1 and NEDD9 signal through SRC to promote E-cadherin removal from the cell membrane and lysosomal degradation. We also find mammary tumors arising in MMTV-polyoma virus T-antigen mice have enhanced junctional E-cadherin in a Nedd9(-/-) background. Cumulatively, these results suggest a new role for Cas proteins in cell-cell adhesion signaling in cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps monoclonal anti-β-actine antibody produced in mouse, clone AC-15, ascites fluid