Merck
FLT1 signaling in metastasis-associated macrophages activates an inflammatory signature that promotes breast cancer metastasis.
The Journal of experimental medicine (2015-08-12)
Bin-Zhi Qian, Hui Zhang, Jiufeng Li, Tianfang He, Eun-Jin Yeo, Daniel Y H Soong, Neil O Carragher, Alison Munro, Alvin Chang, Anne R Bresnick, Richard A Lang, Jeffrey W Pollard
RÉSUMÉ
Although the link between inflammation and cancer initiation is well established, its role in metastatic diseases, the primary cause of cancer deaths, has been poorly explored. Our previous studies identified a population of metastasis-associated macrophages (MAMs) recruited to the lung that promote tumor cell seeding and growth. Here we show that FMS-like tyrosine kinase 1 (Flt1, also known as VEGFR1) labels a subset of macrophages in human breast cancers that are significantly enriched in metastatic sites. In mouse models of breast cancer pulmonary metastasis, MAMs uniquely express FLT1. Using several genetic models, we show that macrophage FLT1 signaling is critical for metastasis. FLT1 inhibition does not affect MAM recruitment to metastatic lesions but regulates a set of inflammatory response genes, including colony-stimulating factor 1 (CSF1), a central regulator of macrophage biology. Using a gain-of-function approach, we show that CSF1-mediated autocrine signaling in MAMs is downstream of FLT1 and can restore the tumor-promoting activity of FLT1-inhibited MAMs. Thus, CSF1 is epistatic to FLT1, establishing a link between FLT1 and inflammatory responses within breast tumor metastases. Importantly, FLT1 inhibition reduces tumor metastatic efficiency even after initial seeding, suggesting that these pathways represent therapeutic targets in metastatic disease.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
trans-2-Bromovinylboronic acid MIDA ester, technical grade