Merck
Effect of nitric acid on pretreatment and fermentation for enhancing ethanol production of rice straw.
Carbohydrate polymers (2013-11-28)
Ilgook Kim, Bomi Lee, Ji-Yeon Park, Sun-A Choi, Jong-In Han
RÉSUMÉ
In this study, nitric acid (HNO₃) was evaluated as an acid catalyst for rice straw pretreatment, and, after neutralization, as a sole nitrogen source for subsequent fermentation. Response surface methodology was used to obtain optimal pretreatment condition with respect to HNO₃ concentration (0.2-1.0%), temperature (120-160 °C) and reaction time (1-20 min). In a condition of 0.65% HNO₃, 158.8 °C and 5.86 min, a maximum xylose yield of 86.5% and an enzymatic digestibility of 83.0% were achieved. The sugar solution that contained nitrate derived from the acid catalyst supported the enhancement of ethanol yield by Pichia stipitis from 10.92 g/L to 14.50 g/L. The results clearly reveal that nitric acid could be used not only as a pretreatment catalyst, but also as a nitrogen source in the fermentation process for bioethanol production. It is anticipated that the HNO₃-based pretreatment can reduce financial burden on the cellulosic bioethanol industry by simplifying after-pretreatment-steps as well as providing a nitrogen source.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acide nitrique, ACS reagent, 70%