- Matrix Gla protein, a new gamma-carboxyglutamic acid-containing protein which is associated with the organic matrix of bone.
Matrix Gla protein, a new gamma-carboxyglutamic acid-containing protein which is associated with the organic matrix of bone.
Biochemical and biophysical research communications (1983-12-28)
P A Price, M R Urist, Y Otawara
PMID6607731
RÉSUMÉ
A new protein has been isolated from CaCl2/urea extracts of demineralized bovine bone matrix. This protein has five to six residues of the vitamin K-dependent amino acid, gamma-carboxyglutamic acid (Gla), and we have accordingly designated it matrix Gla protein. Matrix Gla protein is a 15,000 dalton protein whose amino acid composition includes a single disulfide bond. The absence of 4-hydroxyproline in matrix Gla protein demonstrates that it is not a precursor to bone Gla protein, 5,800 dalton protein which has a residue of 4-hydroxyproline at position 9 in its sequence. Matrix Gla protein also does not cross-react with antibodies raised against bone Gla protein.