Merck
  • Stereotyped subset #1 chronic lymphocytic leukemia: a direct link between B-cell receptor structure, function, and patients' prognosis.
Stereotyped subset #1 chronic lymphocytic leukemia: a direct link between B-cell receptor structure, function, and patients' prognosis.
American journal of hematology (2013-09-14)
Ilaria Del Giudice, Sabina Chiaretti, Simona Santangelo, Simona Tavolaro, Nadia Peragine, Marilisa Marinelli, Caterina Ilari, Sara Raponi, Monica Messina, Mauro Nanni, Francesca Romana Mauro, Alfonso Piciocchi, Katia Bontempi, Davide Rossi, Gianluca Gaidano, Anna Guarini, Robin Foà
RÉSUMÉ
Chronic lymphocytic leukemia (CLL) with stereotyped B-cell receptor (BCR) belonging to subset #1 (IGHV1-5-7/ IGKV1-39) display a poor outcome. To characterize their genetic and genomic features and BCR function, we selected 20 subset #1 CLL from a series of 579 cases. Subset #1 CLL, all showing unmutated IGHV, were associated with the presence of del(11q) (50%) in comparison with unmutated CLL, unmutated stereotyped CLL other than subset #1 and with cases using the same IGHV genes but a heterogeneous VH CDR3 (non-subset #1 CLL). There were no distinctive features regarding CD38, ZAP-70, and TP53 disruption. NOTCH1, SF3B1, and BIRC3 were mutated in 15%, 0%, and 5% of cases, respectively, while BIRC3 was deleted in 22% of cases. Microarray unsupervised analysis on 80 unmutated/mutated/stereotyped/non-stereotyped CLL showed a tight clustering of subset #1 cases. Their genomic signature exhibited several differentially expressed transcripts involved in BCR signal transduction, apoptosis regulation, cell proliferation, and oxidative processes, regardless of del(11q). Accordingly, BCR ligation with anti-IgM revealed a significant higher proliferation of subset #1 versus unmutated non-subset #1 CLL, both at baseline and after 24–48 hr stimulation. Subset #1 CLL represent a paradigmatic example of the direct link between BCR structure, function, and patients prognosis.