Merck
GPRC5B activates obesity-associated inflammatory signaling in adipocytes.
Science signaling (2012-11-22)
Yeon-Jeong Kim, Takamitsu Sano, Takuji Nabetani, Yoshimi Asano, Yoshio Hirabayashi
RÉSUMÉ
A genome-wide association study identified a strong correlation between body mass index and the presence of a 21-kb copy number variation upstream of the human GPRC5B gene; however, the functional role of GPRC5B in obesity remains unknown. We report that GPRC5B-deficient mice were protected from diet-induced obesity and insulin resistance because of reduced inflammation in their white adipose tissue. GPRC5B is a lipid raft-associated transmembrane protein that contains multiple phosphorylated residues in its carboxyl terminus. Phosphorylation of GPRC5B by the tyrosine kinase Fyn and the subsequent direct interaction with Fyn through the Fyn Src homology 2 (SH2) domain were critical for the initiation and progression of inflammatory signaling in adipose tissue. We demonstrated that a GPRC5B mutant lacking the direct binding site for Fyn failed to activate a positive feedback loop of nuclear factor κB-inhibitor of κB kinase ε signaling. These findings suggest that GPRC5B may be a major node in adipose signaling systems linking diet-induced obesity to type 2 diabetes and may open new avenues for therapeutic approaches to diabetic progression.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-phosphotyrosine, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anticorps anti-phosphotyrosine, clone PY20, clone PY20, Upstate®, from mouse