Merck
Synthesis of structurally identical fluorine-18 and iodine isotope labeling compounds for comparative imaging.
Bioconjugate chemistry (2003-03-20)
Zizhong Li, Yu-Shin Ding, Andrew Gifford, Joanna S Fowler, John S Gatley
RÉSUMÉ
The synthesis of a benzophenone-based labeling compound designed for comparative imaging studies with both in vivo positron emission tomograph (PET) and single-photon computed tomography (SPECT) and ex vivo autoradiography is described. The new compound can be labeled with either F-18 or iodine radioisotopes to give two different radioisotopmers: N-[2-fluoro-5-(3-[I-131]iodobenzoyl)benzyl]-2-bromoacetamide (1) and N-[2-[F-18]fluoro-5-(3-iodobenzoyl)benzyl]-2-bromoacetamide (2). Compound 1 and 2 have a 2-bromoacetyl group, which can be used to conjugate with biomolecules through a nucleophilic substitution reaction. Compound 1 was synthesized from the corresponding tributyltin derivatives via an oxidative destannylation reaction, and compound 2 was prepared via a four-step radiosynthesis (nucleophilic aromatic substitution, reduction, oxidation, and alkylation) starting from 4-(N,N,N-trimethylammonio)-3-cyano-3'-iodobenzophenone triflate. A remarkably high radiochemical yield (>90%) was achieved for the F-18 nucleophilic aromatic substitution under mild conditions (room temperature in less than 10 min), indicating the structural advantage of the designed molecule to facilitate the F-18 for trimethylammonium substitution in the presence of two electron-withdrawing groups (nitrile and carbonyl). The overall radiosynthesis time for compound 2 is less than 3 h after end of bombardment (EOB) with an unoptimized radiochemical yield of about 2% (not decay corrected) and specific activity of 0.8 Ci/micromol at EOB. The radiolabeling precursors for compound 1 and 2 were synthesized via a carbon-carbon bond-forming reaction between 2-substituted-5-lithiobenzonitrile and 3-substituted benzaldehyde derivatives. Compounds 1 and 2 should allow us to label biomolecules with F-18 or iodine isotopes and gives structurally identical products, which are expected to have identical biological properties and should be useful for comparative imaging studies.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Bromo-2-fluorobenzonitrile, 97%