Merck
Internal motions prime cIAP1 for rapid activation.
Nature structural & molecular biology (2014-11-11)
Aaron H Phillips, Allyn J Schoeffler, Tsutomu Matsui, Thomas M Weiss, John W Blankenship, Kerry Zobel, Anthony M Giannetti, Erin C Dueber, Wayne J Fairbrother
RÉSUMÉ
Cellular inhibitor of apoptosis 1 (cIAP1) is a ubiquitin ligase with critical roles in the control of programmed cell death and NF-κB signaling. Under normal conditions, the protein exists as an autoinhibited monomer, but proapoptotic signals lead to its dimerization, activation and proteasomal degradation. This view of cIAP1 as a binary switch has been informed by static structural studies that cannot access the protein's dynamics. Here, we use NMR spectroscopy to study micro- and millisecond motions of specific domain interfaces in human cIAP1 and use time-resolved small-angle X-ray scattering to observe the global conformational changes necessary for activation. Although motions within each interface of the 'closed' monomer are insufficient to activate cIAP1, they enable associations with catalytic partners and activation factors. We propose that these internal motions facilitate rapid peptide-induced opening and dimerization of cIAP1, which undergoes a dramatic spring-loaded structural transition.