Merck
PE-1/METS, an antiproliferative Ets repressor factor, is induced by CREB-1/CREM-1 during macrophage differentiation.
The Journal of biological chemistry (2004-02-03)
Dominique Sawka-Verhelle, Laure Escoubet-Lozach, Amy L Fong, Kelly D Hester, Stephan Herzig, Patricia Lebrun, Christopher K Glass
RÉSUMÉ
The molecular mechanisms involved in regulating the balance between cellular proliferation and differentiation remain poorly understood. Members of the Ets-domain family of transcription factors are candidates for proteins that might differentially regulate cell cycle control and cell type-specific genes during the differentiation of myeloid progenitor cells. The Ets repressor PE-1/METS has been suggested to contribute to growth arrest during terminal macrophage differentiation by repressing Ets target genes involved in Ras-dependent proliferation. An important feature of this regulatory model is that PE-1/METS is itself induced by the program of macrophage differentiation elicited by M-CSF. Here, we present evidence that the PE-1/METS gene is a transcriptional target of the cyclic AMP response element-binding protein-1 (CREB-1). CREB-1 expression is dramatically up-regulated during macrophage differentiation and phosphorylation of CREB-1 and the related factor CREM-1 are stimulated by M-CSF in a SAPK2/p38-dependent manner. Chromatin immunoprecipitation experiments demonstrate that CREB-1/CREM-1 are recruited to the PE-1/METS promoter as well as to the promoters of other genes that are up-regulated during terminal macrophage differentiation. Overexpression of CREB-1 stimulates the activities of the PE-1/METS, and macrosialin promoters, while expression of a dominant negative form of CREB-1 during macrophage differentiation inhibits expression of the PE-1/METS and macrosialin genes. Inhibition of CREB function also results in reduced expression of CD54 and impaired cell adhesion. Taken together, these findings reveal new roles of CREB-1/CREM-1 as regulators of macrophage differentiation.