Merck
  • [Synthesis of cytidine diphosphate-3,6-dideoxyhexoses--donors of glycosyl residues in the biosynthesis of O-specific polysaccharides of Salmonella of serogroups A, B and C].
[Synthesis of cytidine diphosphate-3,6-dideoxyhexoses--donors of glycosyl residues in the biosynthesis of O-specific polysaccharides of Salmonella of serogroups A, B and C].
Bioorganicheskaia khimiia (1989-10-01)
N S Utkina, L L Danilov, T N Druzhinina, V N Shibaev
RÉSUMÉ
Interaction of lithium alcoholates of 2,4-di-O-benzoates of paratose and abequose with tetrabenzyl pyrophosphate gave alpha-phosphates of the 3,6-dideoxyhexoses, further converted into the corresponding cytidine-5'-diphosphate derivatives. These synthetic nucleotides were shown to participate in the biosynthesis of the O-specific polysaccharides for Salmonella typhimurium and S. nitra.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Tetrabenzyl pyrophosphate, 98%