Merck
DJ-1( PARK7), a novel gene for autosomal recessive, early onset parkinsonism.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology (2003-11-05)
V Bonifati, P Rizzu, F Squitieri, E Krieger, N Vanacore, J C van Swieten, A Brice, C M van Duijn, B Oostra, G Meco, P Heutink
RÉSUMÉ
Four chromosomal loci ( PARK2, PARK6, PARK7, and PARK9) associated with autosomal recessive, early onset parkinsonism are known. We mapped the PARK7 locus to chromosome 1p36 in a large family from a genetically isolated population in the Netherlands, and confirmed this linkage in an Italian family. By positional cloning within the refined PARK7 critical region we recently identified mutations in the DJ-1 gene in the two PARK7-linked families. The function of DJ-1 remains largely unknown, but evidence from genetic studies on the yeast DJ-1 homologue, and biochemical studies in murine and human cell lines, suggests a role for DJ-1 as an antioxidant and/or a molecular chaperone. Elucidating the role of DJ-1 will lead to a better understanding of the pathogenesis of DJ-1-related and common forms of Parkinson's disease.