Merck
  • Familial hypercatabolic hypoproteinemia caused by deficiency of the neonatal Fc receptor, FcRn, due to a mutant beta2-microglobulin gene.
Familial hypercatabolic hypoproteinemia caused by deficiency of the neonatal Fc receptor, FcRn, due to a mutant beta2-microglobulin gene.
Proceedings of the National Academy of Sciences of the United States of America (2006-03-22)
Manzoor A Wani, Lynn D Haynes, Jonghan Kim, C L Bronson, Chaity Chaudhury, Sudhasri Mohanty, Thomas A Waldmann, John M Robinson, Clark L Anderson
RÉSUMÉ
Two siblings, products of a consanguineous marriage, were markedly deficient in both albumin and IgG because of rapid degradation of these proteins, suggesting a lack of the neonatal Fc receptor, FcRn. FcRn is a heterodimeric receptor composed of a nonclassical MHC class I alpha-chain and beta(2)-microglobulin (beta(2)m) that binds two ligands, IgG and albumin, and extends the catabolic half-lives of both. Eight relatives of the siblings were moderately IgG-deficient. From sera archived for 35 years, we sequenced the two siblings' genes for the heterodimeric FcRn. We found that, although the alpha-chain gene sequences of the siblings were normal, the beta(2)m genes contained a single nucleotide transversion that would mutate a conserved alanine to proline at the midpoint of the signal sequence. Concentrations of soluble beta(2)m and HLA in the siblings' sera were <1% of normal. Transfection assays of beta(2)m-deficient cultured cells with beta(2)m cDNA indicated that the mutant beta(2)m supported <20% of normal expression of beta(2)m, MHC class I, and FcRn proteins. We concluded that a beta(2)m gene mutation underlies the hypercatabolism and reduced serum levels of albumin and IgG in the two siblings with familial hypercatabolic hypoproteinemia. This experiment of nature affirms our hypothesis that FcRn binds IgG and albumin, salvages both from a degradative fate, and maintains their physiologic concentrations.