Merck
From symmetric glycerol derivatives to dissymmetric chlorohydrins.
Molecules (Basel, Switzerland) (2011-03-04)
Carmen Solarte, Marc Escribà, Jordi Eras, Gemma Villorbina, Ramon Canela, Mercè Balcells
RÉSUMÉ
The anticipated worldwide increase in biodiesel production will result in an accumulation of glycerol for which there are insufficient conventional uses. The surplus of this by-product has increased rapidly during the last decade, prompting a search for new glycerol applications. We describe here the synthesis of dissymmetric chlorohydrin esters from symmetric 1,3-dichloro-2-propyl esters obtained from glycerol. We studied the influence of two solvents: 1,4-dioxane and 1-butanol and two bases: sodium carbonate and 1-butylimidazole, on the synthesis of dissymmetric chlorohydrin esters. In addition, we studied the influence of other bases (potassium and lithium carbonates) in the reaction using 1,4-dioxane as the solvent. The highest yield was obtained using 1,4-dioxane and sodium carbonate.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Butylimidazole, 98%