Merck
Structure and chromosomal localization of the human 2',3'-cyclic nucleotide 3'-phosphodiesterase gene.
Annals of human genetics (1992-07-01)
A J Douglas, M F Fox, C M Abbott, L J Hinks, G Sharpe, S Povey, R J Thompson
RÉSUMÉ
Human brain cDNA clones for the myelin associated enzyme 2',3'-cyclic nucleotide 3'-phosphodiesterase (CNPase) have been isolated and sequenced. The only 5' untranslated region (UTR) sequence found was that of a human CNPII mRNA, with no direct evidence for a CNPI mRNA. Human CNPase cDNAs were used to isolate genomic clones containing the human CNPase gene which is 9 kb long. Four exons were identified, separated by three introns, and the sequence of each exon and intron/exon boundary has been established. The polymerase chain reaction (PCR) was used to detect the presence of the human CNPase gene in DNA from a panel of rodent/human somatic cell hybrids. By this means the human CNPase gene was mapped to chromosome 17. In situ hybridization of a human CNPase genomic clone to metaphase chromosomes further localized this gene to chromosomal band 17q21.