Merck
Vibrational spectroscopic studies of mesna and dimesna.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-05-09)
Ying-Sing Li, Yu Wang, Jeffrey S Church, Fabio Garzena, Zanxing Zhang, Danqui An
RÉSUMÉ
Raman, and infrared spectra of mesna and dimesna have been collected in the present spectroscopic studies. Based on the group frequencies, relative intensities and Raman depolarization measurements, some vibrational assignments have been suggested. For both mesna and dimesna, at least two rotational conformers have been identified. Adsorption behavior was investigated from the recorded surface-enhanced Raman scattering (SERS) spectra. It was found that both mesna and dimesna adsorbed as thiolate on silver sol particles with the cleavage of the S-H bond in mesna and the S-S bond in dimesna. For the adsorbed thiolate, two conformers existed in the adsorption state.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Mesna impurity D, European Pharmacopoeia (EP) Reference Standard