Merck
  • A simple method for estimating activation energies using the fragmentation yield: collision-induced dissociation of iron(II)-phenanthroline complexes in an electrospray ionization mass spectrometer.
A simple method for estimating activation energies using the fragmentation yield: collision-induced dissociation of iron(II)-phenanthroline complexes in an electrospray ionization mass spectrometer.
Journal of the American Society for Mass Spectrometry (2006-05-23)
Sandor Keki, Lajos Nagy, Janos Torok, Gyorgy Deak, Miklos Zsuga
RÉSUMÉ
The gas-phase stabilities of Fe(Phi)3(2+) complexes, where Phi represents the 1,10-phenanthroline, 5-chloro-1,10-phenanthroline, 5-methyl-1,10-phenanthroline, 3,4,7,8-tetramethyl-1,10-phenanthroline, and 4,7-diphenyl-1,10-phenanthroline ligands were investigated by collision-induced dissociation (CID) in the capillary-first skimmer region upon changing the voltage difference between the capillary and the skimmer. The loss of only one ligand from the Fe(Phi)3(2+) complexes was observed with each of the phenanthroline ligands studied. An increase in the voltage difference between the capillary and the skimmer resulted in a higher fragmentation yield as calculated from the intensity of the precursor and the fragment ion. The fragmentation yield versus capillary-skimmer voltage difference plots were evaluated by means of the Arrhenius and the Rice-Ramsperger-Kassel (RRK) model by fitting the model parameters to the experimental data. Both models yielded practically the same results. In addition, if the internal energy gained through the capillary-skimmer region is estimated correctly, the approximate value of the critical energy (activation energy) for fragmentation can be extracted from the fragmentation yield versus capillary-skimmer voltage difference plots. It was found that the gas-phase stabilities of the Fe(Phi)3(2+) complexes are nearly identical except for the more stable Fe(II)-4,7-diphenyl-1,10-phenanthroline complex. The critical energy for fragmentation was estimated to be approximately 1.2 and 0.9 eV for the Fe(II)- 4,7-diphenyl-1,10-phenanthroline, and the other complexes, respectively.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Methyl-1,10-phenanthroline, ≥99%