Merck
Ultrafast excited-state proton-transfer reaction of 1-naphthol-3,6-disulfonate and several 5-substituted 1-naphthol derivatives.
The journal of physical chemistry. B (2013-01-12)
Mirabelle Prémont-Schwarz, Tamar Barak, Dina Pines, Erik T J Nibbering, Ehud Pines
RÉSUMÉ
The 1-naphthol molecule has been the subject of intense research activity for the past 60 years due to its complex behavior as a photoacid upon optical excitation. We have utilized femtosecond mid-infrared spectroscopy and time-resolved fluorescence spectroscopy to investigate the excited-state proton-transfer reaction of 1-naphthol-3,6-disulfonate (1N-3,6diS) and several 5-substituted 1-naphthol derivatives. The proton dissociation rate constant of 1N-3,6-diS was found to be about 3 times faster and the pKa* about 2 pKa units more acidic than the values previously reported in the literature. A Marcus (free-energy) plot of excited-state proton dissociation rate constants vs the excited-state equilibrium constant of the photoacids, Ka*, was constructed using the C-5 series of 1-naphthol derivatives. The newly measured values for the ESPT rate constant and pKa* of 1N-3,6diS was found to fit well with the Marcus correlation. We discuss our findings in the context of the photoacidity phenomenon in general, and the photoacidity of 1-naphthol and its derivatives in particular.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Naphthol-3,6-disulfonic acid disodium salt hydrate, technical grade