Merck
  • The assay on a defined medium of the effects of beta-2-thienylalanine on the growth of anaerobic bacterial isolates from phenylketonuric patients.
The assay on a defined medium of the effects of beta-2-thienylalanine on the growth of anaerobic bacterial isolates from phenylketonuric patients.
Medical microbiology and immunology (1980-02-01)
K J Brown, B V Vesey, G W Tannock, E B Bell, D R Lines, R B Elliott
RÉSUMÉ
Faecal samples were taken from three diet-managed phenylketonuric children to determine effects of beta-2-thienylalanine (beta-2-t) on indigenous bacteria. From sample swabs, 127 anaerobes were identified and tested for beta-2-t inhibition on a phenylalanine (Phe)-free medium, Anaerobe Inhibition Test (AIT) agar. Of the isolates, 77.9% grew sufficiently to assay reactions on at least 25% of AIT plates. Using Phe-containing Columbia agar, 86.5% of the strains could be assayed. None of 28 Bacteroides cultures was inhibited by beta-2-t on AIT. Of the genera, Bifidobacterium, Eubacterium, Lactobacillus, Peptostreptococcus, and Propionibacterium, no isolates which would grow on AIT were inhibited. At least one isolate of each of the genera Peptococcus, Fusobacterium, and Clostridium was inhibited. Of 127 total isolates, only nine were inhibited by beta-2-t on AIT, and inhibition was abolished on Columbia agar. Thirty-nine "aerobes" were isolated from the same patients. Strains of the genera tested reacted similarly to previously tested strains from non-PKU sources. Also, anaerobically isolated Excherichia coli were inhibited, while Streptococcus faecalis cultures were not, confirming results on aerobically-isolated non-PKU cultures of the same species. These studies, the first dealing with beta-2-t and anaerobic bacteria, suggest that little change in intestinal bacterial populations might be expected during in vivo beta-2-t treatment.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-(2-Thienyl)-L-alanine, ≥98.0% (TLC)
Sigma-Aldrich
3-(2-Thienyl)-DL-alanine, ≥98%