Merck
Syntheses and detailed structure characterization of dextran carbonates.
Carbohydrate polymers (2013-03-08)
Thomas Elschner, Holger Wondraczek, Thomas Heinze
RÉSUMÉ
Dextran alkyl carbonates were synthesized applying ethyl chloroformate, butyl chloroformate, butyl fluoroformate and 1H-imidazole-1-carboxylates. The influence of the reaction conditions on the reaction efficiency and the substitution pattern was studied in detail. The structure of the products obtained was clearly described by means of NMR- and IR-spectroscopy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ergosterol, ≥75%
Sigma-Aldrich
Butyl chloroformate, 98%