Merck
A stereoselective cyclization strategy for the preparation of γ-lactams and their use in the synthesis of α-methyl-β-proline.
The Journal of organic chemistry (2012-11-07)
Souvik Banerjee, Justin Smith, Jillian Smith, Caleb Faulkner, Douglas S Masterson
RÉSUMÉ
A straightforward stereoselective and enantiodivergent cyclization strategy for the construction of γ-lactams is described. The cyclization strategy makes use of chiral malonic esters prepared from enantiomerically enriched monoesters of disubstituted malonic acid. The cyclization occurs with the selective displacement of a substituted benzyl alcohol as the leaving group. A Hammett study illustrates that the cyclization is under electronic control. The resulting γ-lactam can be readily converted into a novel proline analogue.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)