Merck
Chemical constituents of Bauhinia aurea.
Journal of Asian natural products research (2012-10-11)
Xiao-Ya Shang, Shuai Li, Su-Juan Wang, Yong-Chun Yang, Jian-Gong Shi
RÉSUMÉ
A new dihydroflavonol glycoside dimer 6,6-bisastilbin (1) and a new nitrile-containing metabolite (Z)-5α,6β-dihydroxy-4β-methoxy-2-cyclohexene-Δ(1,α)-acetonitrile (2), together with three known analogs, bauhinin, bauhinilide, and dehydrodicatechin A, have been isolated from an ethanol extract of Bauhinia aurea. Their structures were determined by spectroscopic and chemical methods.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Astilbin from Engelhardtia roxburghiana, ≥98%