Merck
Measurable urinary albumin predicts cardiovascular risk among normoalbuminuric patients with type 2 diabetes.
Journal of the American Society of Nephrology : JASN (2012-09-01)
Piero Ruggenenti, Esteban Porrini, Nicola Motterlini, Annalisa Perna, Aneliya Parvanova Ilieva, Ilian Petrov Iliev, Alessandro Roberto Dodesini, Roberto Trevisan, Antonio Bossi, Giuseppe Sampietro, Enrica Capitoni, Flavio Gaspari, Nadia Rubis, Bogdan Ene-Iordache, Giuseppe Remuzzi
RÉSUMÉ
Micro- or macroalbuminuria is associated with increased cardiovascular risk factors among patients with type 2 diabetes, but whether albuminuria within the normal range predicts long-term cardiovascular risk is unknown. We evaluated the relationships between albuminuria and cardiovascular events in 1208 hypertensive, normoalbuminuric patients with type 2 diabetes from the BErgamo NEphrologic Diabetes Complication Trial (BENEDICT), all of whom received angiotensin-converting enzyme inhibitor (ACEI) therapy at the end of the trial and were followed for a median of 9.2 years. The main outcome was time to the first of fatal or nonfatal myocardial infarction; stroke; coronary, carotid, or peripheral artery revascularization; or hospitalization for heart failure. Overall, 189 (15.6%) of the patients experienced a main outcome event (2.14 events/100 patient-years); 24 events were fatal. Albuminuria independently predicted events (hazard ratio [HR], 1.05; 95% confidence interval [CI], 1.02-1.08). Second-degree polynomial multivariable analysis showed a continuous nonlinear relationship between albuminuria and events without thresholds. Considering the entire study population, even albuminuria at 1-2 μg/min was significantly associated with increased risk compared with albuminuria <1 μg/min (HR, 1.04; 95% CI, 1.02-1.07). This relationship was similar in the subgroup originally randomly assigned to non-ACEI therapy. Among those originally receiving ACEI therapy, however, the event rate was uniformly low and was not significantly associated with albuminuria. Taken together, among normoalbuminuric patients with type 2 diabetes, any degree of measurable albuminuria bears significant cardiovascular risk. The association with risk is continuous but is lost with early ACEI therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder