Merck
  • Quantitation of hypoxanthine in plasma from patients with ischemic heart disease: adaption of a high-performance liquid chromatographic method.
Quantitation of hypoxanthine in plasma from patients with ischemic heart disease: adaption of a high-performance liquid chromatographic method.
Journal of chromatography (1991-07-17)
H M Himmel, V Sadony, U Ravens
RÉSUMÉ
A high-performance liquid chromatographic method is described for the separation and quantitation of several purine compounds, including hypoxanthine. The isocratic separation of a standard mixture of nine compounds is achieved within 20 min on a reversed-phase Nucleosil 100-5C18 column, with a mobile phase of KH2PO4 (300 mM, pH 4.0)-methanol-acetonitrile-tetrahydrofuran (97.9:1:1:0.1, v/v). Uric acid, guanine, hypoxanthine, uridine, xanthine, allopurinol, inosine, guanosine and 7-methylxanthine were almost completely baseline-separated, with detection limits in the range 0.5-1.2 pmol per injection. The influence of the concentrations of buffer and tetrahydrofuran on the quality of separation are described. The within-day and the day-to-day precision were satisfactory (e.g. coefficients of variation of less than 1.5 and ca. 6.0%, respectively, for peak heights). The recovery of [3H]hypoxanthine added to samples was 86 +/- 1%. Hypoxanthine was quantified in human plasma samples obtained at various times during coronary artery bypass grafting. The hypoxanthine levels measured immediately after release of the aortic cross-clamp were significantly higher than those determined under control conditions (18.8 +/- 7.0 and 3.4 +/- 1.0 microM, respectively).