Merck
  • Metabolism of 3-chloro-4-fluoroaniline in rat using [14C]-radiolabelling, 19F-NMR spectroscopy, HPLC-MS/MS, HPLC-ICPMS and HPLC-NMR.
Metabolism of 3-chloro-4-fluoroaniline in rat using [14C]-radiolabelling, 19F-NMR spectroscopy, HPLC-MS/MS, HPLC-ICPMS and HPLC-NMR.
Xenobiotica; the fate of foreign compounds in biological systems (2006-03-02)
C J Duckett, J C Lindon, H Walker, F Abou-Shakra, I D Wilson, J K Nicholson
RÉSUMÉ
The metabolic fate of 3-chloro-4-fluoroaniline was investigated in rat following intraperitoneal (i.p.) administration at 5 and 50 mg kg(-1) using a combination of HPLC-MS, HPLC-MS/MS, (19)F-NMR spectroscopy, HPLC-NMR spectroscopy and high-pressure liquid chromatography-inductively coupled plasma mass spectrometry (HPLC-ICPMS) with (35)Cl and (34)S detection. The metabolism of 3-chloro-4-fluoroaniline at both doses was rapid and extensive, to a large number of metabolites, with little unchanged compound excreted via the urine. Dosing at 5 mg kg(-1) with [(14)C]-labelled compound enabled the comparison of standard radioassay analysis methods with (19)F-NMR spectroscopy. (19)F-NMR resonances were only readily detectable in the 0-12 h post-dose samples. Dosing at 50 mg kg(-1) allowed the facile and specific detection and quantification of metabolites by (19)F-NMR spectroscopy. Metabolite profiling was also possible at this dose level using HPLC-ICPMS with (35)Cl-specific detection. The principal metabolites of 3-chloro-4-fluoroaniline were identified as 2-amino-4-chloro-5-fluorophenyl sulfate and 2-acetamido-4-chloro-5-fluorophenyl glucuronide. N-acetylation and hydroxylation followed by O-sulfation were the major metabolic transformations observed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Chloro-4-fluoroaniline, 98%