Merck
Neurokinin-1 receptor in peripheral nerve terminals mediates thermal hyperalgesia.
Biochemical and biophysical research communications (2005-11-23)
Wen-Ling Chen, Yu-Qiu Zhang, Zhi-Qi Zhao
RÉSUMÉ
Neurokinin-1 receptor (NK-1) plays an important role in nociception. The present study was to explore whether activation of peripheral NK-1 receptor, especially expressed on primary sensory afferents, could induce hyperalgesia and sensitize C-type sensory afferents. (1) Intraplantar administration of NK-1 agonist [Sar9, Met(O2)11]SP (Sar-SP, 0.2, 1 nmol, 20 microl) produced significant thermal hyperalgesia and edema, which was blocked by co-injection of NK-1 antagonist WIN51,708 (10 nmol). But in the rats with compound 48/80 treatment for mast cell depletion, the Sar-SP-induced edema, but not hyperalgesia, was attenuated. (2) Close-arterial injection of Sar-SP (1 nmol, 0.1 ml) excited and sensitized sensory C afferents of the sural nerve to heat stimuli. The results suggest involvement of NK-1 receptors expressed on the peripheral afferent terminals in thermal hyperalgesia mediated by directly sensitizing C-type sensory afferents.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
[Sar9, Met(O2)11]-Substance P, ≥95% (HPLC)