Merck
Properties and applications of the (2-nitrofluoren-9-yl)methoxycarbonyl group.
Journal of peptide science : an official publication of the European Peptide Society (2001-04-12)
B Henkel, E Bayer
RÉSUMÉ
This paper presents a new protecting group, the (2-nitrofluoren-9-yl)methoxycarbonyl group. Investigations on the properties of this new modification of the Fmoc-system, such as the solvent-dependent photochemical cleavage, and enhanced lability towards bases, are described, as well as UV-kinetic measurements of the cleavage reaction. In addition, the incorporation of the (2-nitrofluoren-9-yl)methoxycarbonyl group into two peptides, and a sequence-dependent photochemical cleavage reaction are reported.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Nitro-9-fluorenone, 99%