Merck
Agonists with supraphysiological efficacy at the muscarinic M2 ACh receptor.
British journal of pharmacology (2012-10-16)
R Schrage, W K Seemann, J Klöckner, C Dallanoce, K Racké, E Kostenis, M De Amici, U Holzgrabe, K Mohr
RÉSUMÉ
Artificial agonists may have higher efficacy for receptor activation than the physiological agonist. Until now, such 'superagonism' has rarely been reported for GPCRs. Iperoxo is an extremely potent muscarinic receptor agonist. We hypothesized that iperoxo is a 'superagonist'. Signalling of iperoxo and newly synthesized structural analogues was compared with that of ACh at label-free M2 muscarinic receptors applying whole cell dynamic mass redistribution, measurement of G-protein activation, evaluation of cell surface agonist binding and computation of operational efficacies. In CHO-hM2 cells, iperoxo significantly exceeds ACh in Gi /Gs signalling competence. In the orthosteric loss-of-function mutant M2 -Y104(3.33) A, the maximum effect of iperoxo is hardly compromised in contrast to ACh. 'Superagonism' is preserved in the physiological cellular context of MRC-5 human lung fibroblasts. Structure-signalling relationships including iperoxo derivatives with either modified positively charged head group or altered tail suggest that 'superagonism' of iperoxo is mechanistically based on parallel activation of the receptor protein via two orthosteric interaction points. Supraphysiological agonist efficacy at muscarinic M2 ACh receptors is demonstrated for the first time. In addition, a possible underlying molecular mechanism of GPCR 'superagonism' is provided. We suggest that iperoxo-like orthosteric GPCR activation is a new avenue towards a novel class of receptor activators.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Iperoxo, ≥98% (HPLC)