Merck
Renal drug targeting using a vector "alkylglycoside".
The Journal of pharmacology and experimental therapeutics (1998-12-23)
K Suzuki, H Susaki, S Okuno, Y Sugiyama
RÉSUMÉ
A specific sugar-modified peptide has previously been shown to have renal targeting potential in vivo and to have a specific binding site which has been identified in the kidney membrane fraction. In this report, we studied the inhibitory effects of glycosylated derivatives on the binding of [3H]Glc-O-C8-AVP [a glucosylated derivative of Arg8-vasopressin (AVP), Kd = 55 nM] to clarify the structural requirements necessary for renal recognition. Glc-S-C7-Me (octyl beta-D-thioglucoside) markedly inhibited the binding, to a much greater extent than Glc-O-C7-Me (octyl beta-D-glucoside) and Gal-S-C7-Me (octyl beta-D-thiogalactoside). Also, [3H]Glc-S-C7-Me was shown to have a specific binding site on the kidney membrane (Kd = 17 nM, Bmax = 24 pmol/mg protein) rather than the liver membrane and, in addition, Glc-S-C7-Me exhibited effective and selective renal uptake in vivo. To examine the possibility that Glc-S-C7-Me might be of practical use as a renal targeting vector, AVP, tryptamine and 4-nitrobenz-2-oxa-1,3-diazole were modified with Glc-S-C8- and the tissue uptake of the resulting derivatives was evaluated. All of these derivatives showed clear renal targeting potential because the apparent uptake clearance by the kidney was greater than 3 ml/min/g kidney in each case. As far as the AVP derivatives were concerned, derivatives having different numbers of methylene groups were compared with Glc-S-C8-AVP. Glc-S-C11-AVP exhibited increased kidney targeting potential, whereas that of Glc-S-C5-AVP was reduced. These differences suggest that the "alkylglycoside" moiety is important for renal uptake. In addition, these renally targeted derivatives inhibited the binding of [3H]Glc-S-C7-Me to the kidney membrane fraction. Our findings allow us to conclude that the alkylglycoside is a suitable candidate vector for renal targeting.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)