Merck
[Estimation of pathways of 5-fluorouracil anabolism in human cancer cells in vitro and in vivo].
Gan to kagaku ryoho. Cancer & chemotherapy (1996-05-01)
M Fukushima, H Nomura, Y Murakami, T Shirasaka, K Aiba
RÉSUMÉ
Possible pathways of intracellular phosphorylation of 5-fluorouracil (5-FU) in human cancer cells were investigated in vitro and in vivo. We used two inhibitors which regulate the anabolism of 5-FU for the purpose of elucidation of its pathways; one is oxonic acid (Oxo), an inhibitor of orotate phosphoribosyltransferase (OPRTase), catalizing a formation of FUMP from 5-FU, and another is 2, 6-dihydroxypyridine (DP), an inhibitor of uracil ribosyltransferase which catalizes a conversion of 5-FU to 5-fluorouridine. Although the pathway of 5-FU phosphorylation in murine tumor cells was varied, about 80% of human cancer cells tested were found to utilize the first pathway in which 5-FU was converted to FUMP by OPRTase. Thus, the phosphorylation of 5-FU via the first pathway was markedly inhibited by Oxo in 4 strains of 5 gastric cancer cells, 7 of 8 colorectal cancer cells and 3 of 4 lung cancer cells. In xenografts of human gastric and colorectal adenocarcinoma in which 5-FU phosphorylation is regulated by Oxo in vitro, the production of 5-fluoronucleotides and its incorporation into RNA after iv administration of 5-FU significantly decreased by co-administration of Oxo, suggesting that the nature of an anabolic pathway of 5-FU in the tumor cells in vitro reflects in vivo behavior. We also confirmed that the phosphoribosylpyrophosphate level in tumor cells was importantly related to determination of the metabolic pathway of 5-FU. These results would suggest that possible modulation of 5-FU lies on the augmentation of 5-FU efficacy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,6-Dihydroxypyridine hydrochloride, 97%