Merck
Characterization of a specific alpha-mannosidase involved in oligosaccharide processing in Saccharomyces cerevisiae.
The Journal of biological chemistry (1985-02-25)
S Jelinek-Kelly, T Akiyama, B Saunier, J S Tkacz, A Herscovics
RÉSUMÉ
Fractionation of a crude extract from Saccharomyces cerevisiae X-2180 on Sepharose 6B in the presence of 0.5% Triton X-100 resolves two enzyme fractions containing alpha-mannosidase activity. Fraction I which is excluded from the gel contains alpha-mannosidase activity toward both p-nitrophenyl-alpha-D-mannopyranoside and Man9GlcNAc oligosaccharide as substrates, whereas Fraction II which is included in the gel contains only oligosaccharide alpha-mannosidase activity. The latter enzyme is very specific and removes a single mannose residue from Man9GlcNAc, whereas the alpha-mannosidase activity of Fraction I removes several mannose residues from Man9GlcNAc oligosaccharide. High resolution 1H NMR analysis of the Man8GlcNAc formed from Man9GlcNAc in the presence of the alpha-mannosidase of Fraction II showed only a single isomer with the following structure: (see formula; see text) This specific enzyme is most probably involved in processing of oligosaccharide during biosynthesis of mannoproteins. The mannose analog of 1-deoxynojirimycin (50-500 microM), dideoxy-1,5-imino-D-mannitol, inhibits the oligosaccharide alpha-mannosidase activities of Fractions I and II to about the same extent, but has no effect on the nonspecific alpha-mannosidase which acts on p-nitrophenyl-alpha-D-mannopyranoside.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Nitrophenyl α-D-mannopyranoside, α-mannosidase substrate