Merck
Synthesis and radiosensitizing properties of some 2-furancarboxylic acid derivatives.
Polish journal of pharmacology and pharmacy (1985-09-01)
J Tułecki, J Dudzińska-Usarewicz, P Radoła
RÉSUMÉ
New S-2-furoyl-O-alkyldithiocarbonic anhydrides have been obtained by condensation of 2-furoyl chloride with potassium O-alkyl-dithiocarbonates. The toxicity and radiosensitizing activity of these compounds were determined.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Furoyl chloride, 95%