Merck
Indices used in differentiation of thalassemia trait from iron deficiency anemia in pediatric population: are they reliable?
Pediatric hematology and oncology (2012-08-08)
Burçin Nalbantoğlu, Savaş Güzel, Volkan Büyükyalçın, M Metin Donma, Eda Çelik Güzel, Ayşin Nalbantoğlu, Erkut Karasu, Burcu Özdilek
RÉSUMÉ
Iron deficiency (IDA) and beta thalassemia trait (TT) are the most common causes of hypochromia and microcytosis. Many indices have been defined to quickly discriminate these similar entities via parameters obtained from automated blood cell analyzers. However, studies in the pediatric age group are scarce and their results are controversial. We calculated eight discrimination indices [Mentzer Index (MI), England and Fraser Index (E&F), Srivastava Index (S), Green and King Index (G&K), Shine and Lal Index (S&L), red blood cell (RBC) count, RBC distribution width, and red blood cell distribution width Index (RDWI)] in 100 patients. We calculated sensitivity (SENS), specificity (SPEC), positive and negative predictive value (PPV and NPV), and Youden's Index (YI) of each discrimination index. None of the discrimination indices showed a SENS and SPEC of 100%. The highest SENS was obtained with S&L (87.1%), while the highest SPEC was obtained with E&F formula (100%). The highest YI value was obtained with E&F formula (58.1%). In our study, none of the formulas appears reliable in discriminating between TT and IDA patients. The evaluation of iron status and measurement of hemoglobin A(2) (HbA(2)) remain the most reliable investigations to differentiate between TT and IDA patients.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Hemoglobin A2, Ferrous Stabilized human, lyophilized powder