Merck
Wake promoting agents: search for next generation modafinil, lessons learned: part III.
Bioorganic & medicinal chemistry letters (2012-05-02)
Derek Dunn, Greg Hostetler, Mohamed Iqbal, Val R Marcy, Yin Guo Lin, Bruce Jones, Lisa D Aimone, John Gruner, Mark A Ator, Edward R Bacon, Sankar Chatterjee
RÉSUMÉ
In searching for a next generation molecule to the novel wake promoting agent modafinil (compound 1), a series of fluorene-derived wakefulness enhancing agents were developed and evaluated in rat. Extensive pharmacokinetic studies of a potent member of the series (compound 15) revealed that the wake promotion activity of the analog was likely due to an active metabolite (compound 3).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
9-Hydroxyfluorene, 96%