Merck
Adenosine triphosphate-sensitive potassium channel blockers attenuate the antiallodynic effect of R-PIA in neuropathic rats.
Anesthesia and analgesia (2011-05-06)
Jun-Gol Song, Kyung Don Hahm, Young Ki Kim, Jeong Gil Leem, Chung Lee, Sung Moon Jeong, Pyung Hwan Park, Jin Woo Shin
RÉSUMÉ
Nerve injury can generate neuropathic pain. The accompanying mechanical allodynia may be reduced by the intrathecal administration of adenosine. The neuroprotective effects of adenosine are mediated by the adenosine triphosphate (ATP)-sensitive potassium (K(ATP)) channel. We assessed the relationship between the adenosine A1 receptor agonist, N⁶-(R)-phenylisopropyl adenosine (R-PIA), and K(ATP) channels to determine whether the antiallodynic effects of R-PIA are also mediated through K(ATP) channels in a rat nerve ligation injury model of neuropathic pain. Mechanical allodynia was induced by tight ligation of the left lumbar fifth and sixth spinal nerves. Mechanical allodynia in the left hindpaw was evaluated using von Frey filaments to measure withdrawal thresholds. R-PIA (0.5, 1, or 2 μg) was administered intrathecally to induce antiallodynia. We assessed whether pretreatment with the K(ATP) channel blockers glibenclamide or 5-hydroxydecanoate reversed the antiallodynic effect of R-PIA. Also, we evaluated whether diazoxide, a K(ATP) channel opener, had an antiallodynic effect and promoted the antiallodynic effect of R-PIA. Lastly, we investigated whether the voltage-activated K channel blocker 4-aminopyridine attenuated the effect of R-PIA. Intrathecal R-PIA produced maximal antiallodynia at 2 μg (P < 0.05). Intrathecal pretreatment with glibenclamide and intraperitoneal pretreatment 5-hydroxydecanoate significantly reduced the antiallodynic effect of R-PIA. Diazoxide produced an antiallodynic effect and also enhanced the antiallodynic action of R-PIA. 4-Aminopyridine had no effect on the antiallodynic action of R-PIA. The antiallodynic effects of adenosine A1 receptor stimulation may be related to K(ATP) channel activity in a rat model of nerve ligation injury.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(−)-N6-(2-Phenylisopropyl)adenosine, solid