Merck
Complete NMR data of methoxylated cis- and trans-stilbenes as well as 1,2-diphenylethanes.
Magnetic resonance in chemistry : MRC (2011-04-01)
Geunhyeong Jo, Jiye Hyun, Doseok Hwang, Young Han Lee, Dongsoo Koh, Yoongho Lim
RÉSUMÉ
Resveratrol is a polyphenol isolated from many natural sources including grapes, mulberries, eucalyptus, spruce, lilies, and peanuts. The hydroxyl groups in polyphenols can be substituted with various functional groups, allowing production of multiple derivatives. NMR spectroscopy is used to identify new derivatives. Since the complete NMR data of the known derivatives can be useful for identification of the newly isolated derivatives, here, we report the synthesis of 14 methoxylated stilbenes and four 1,2-diphenylethanes and their NMR data.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Bibenzyl, ReagentPlus®, 99%