Merck
A powerful synergistic effect for highly efficient diastereo- and enantioselective phase-transfer catalyzed conjugate additions.
Chemical communications (Cambridge, England) (2010-12-01)
Ming-Qing Hua, Lian Wang, Han-Feng Cui, Jing Nie, Xiao-Ling Zhang, Jun-An Ma
RÉSUMÉ
An efficient, catalytic, diastereo- and enantioselective conjugate addition of N-(diphenylmethylene)glycine tert-butyl ester to β-aryl substituted enones was realized in the presence of 1 mol% of newly desired dinuclear N-spiro-ammonium salts, affording functionalized α-amino acid derivatives in 57-98% yields with high diastereoselectivity (up to 99:1 dr) and enantioselectivity (up to 96.5:3.5 er).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Glycine tert-butyl ester hydrochloride, 97%