Merck
Validation of a liquid chromatography method for the simultaneous quantification of ochratoxin A and its analogues in red wines.
Journal of chromatography. A (2010-11-30)
Rebeca Remiro, María Ibáñez-Vea, Elena González-Peñas, Elena Lizarraga
RÉSUMÉ
A validated high-performance liquid chromatography (HPLC) method with fluorescence detection for the simultaneous quantification of ochratoxin A (OTA) and its analogues (ochratoxin B (OTB), ochratoxin C (OTC) and methyl ochratoxin A (MeOTA)) in red wine at trace levels is described. Before their analysis by HPLC-FLD, ochratoxins were extracted and purified with immunoaffinity columns from 50 mL of red wine at pH 7.2. Validation of the analytical method was based on the following parameters: selectivity, linearity, robustness, limits of detection and quantification, precision (within-day and between-day variability), recovery and stability. The limits of detection (LOD) in red wine were established at 0.16, 0.32, 0.27 and 0.17 ng L(-1) for OTA, OTB, MeOTA and OTC, respectively. The limit of quantification (LOQ) was established as 0.50 ng L(-1) for all of the ochratoxins. The LOD and LOQ obtained are the lowest found for OTA in the reference literature up to now. Recovery values were 93.5, 81.7, 76.0 and 73.4% for OTA, OTB, MeOTA and OTC, respectively. For the first time, this validated method permits the investigation of the co-occurrence of ochratoxins A, B, C and methyl ochratoxin A in 20 red wine samples from Spain.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Ochratoxine B solution, ~10 μg/mL in acetonitrile, analytical standard