Merck
Simple and facile synthesis of tetralone-spiro-glutarimides and spiro-bisglutarimides from Baylis-Hillman acetates.
Organic & biomolecular chemistry (2008-03-11)
Deevi Basavaiah, Raju Jannapu Reddy
RÉSUMÉ
A simple and convenient synthesis of di(E)-arylidene-tetralone-spiro-glutarimides from Baylis-Hillman acetates via an interesting biscyclization strategy involving facile C-C and C-N bond formation is described. Also, one-pot multistep transformation of the Baylis-Hillman acetates into di(E)-arylidene-spiro-bisglutarimides is presented.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Glutarimide, 98%