Merck
Toward chiral sum-frequency spectroscopy.
Journal of the American Chemical Society (2006-07-06)
Na Ji, Victor Ostroverkhov, Mikhail Belkin, Ying-Jen Shiu, Yuen-Ron Shen
RÉSUMÉ
Chiral sum-frequency (SF) spectroscopy that measures both the real and the imaginary components of the SF spectral response was demonstrated for the first time. It was based on interference of the SF signal with a dispersionless SF reference. Solutions of 1,1'-bi-2-naphthol (BN) were used as model systems, and their chiral SF spectra over the first exciton-split transitions were obtained. Chiral spectra are useful for determination of absolute configuration and conformation of chiral molecules.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,1′-Bi-2-naphthol, 99%