Merck
Facile synthesis of highly substituted 3-aminofurans from thiazolium salts, aldehydes, and dimethyl acetylenedicarboxylate.
The Journal of organic chemistry (2005-10-22)
Cheng Ma, Hanfeng Ding, Guangming Wu, Yewei Yang
RÉSUMÉ
[reaction: see text] A facile preparation of 3-aminofuran derivatives via multicomponent reactions of thiazole carbenes, aldehydes, and dimethyl acetylenedicarboxylate (DMAD) is reported. In this process, the thiazole carbenes, generated in situ from thiazolium salts, reacted with aldehydes and DMAD at -78 to 0 degree C in CH(2)Cl(2) to afford the substituted furans in moderate to good yields. Eight substituted thiazolium salts were employed as carbene precursors in the reaction. Besides aryl aldehydes, alpha,beta-unsaturated aldehydes, aliphatic aldehydes, and arenedial were also investigated and found to be applicable to this reaction.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%