- Probing the structure of a rotaxane with two-dimensional infrared spectroscopy.
Probing the structure of a rotaxane with two-dimensional infrared spectroscopy.
Proceedings of the National Academy of Sciences of the United States of America (2005-09-10)
Olaf F A Larsen, Pavol Bodis, Wybren Jan Buma, Jeffrey S Hannam, David A Leigh, Sander Woutersen
PMID16150716
RÉSUMÉ
Femtosecond 2D-IR spectroscopy has been used to study the structure of a [2]rotaxane composed of a benzylic amide macrocycle that is mechanically interlocked onto a succinamide-based thread. Both the macrocycle and the thread contain carbonyl groups, and by determining the coupling between the stretching modes of these groups from the cross-peaks in the 2D-IR spectrum, the structure of the macrocycle-thread system has been probed. Our results demonstrate that 2D-IR spectroscopy can be used to observe structural changes in molecular devices on a picosecond time scale.
MATÉRIAUX