Merck
1,25(OH)2D3 blunts hormone-elevated cytosolic Ca2+ in osteoblast-like cells.
The American journal of physiology (1992-12-01)
J Green, C R Kleeman, S Schotland, L H Ye
RÉSUMÉ
Cytosolic free calcium ([Ca2+]i) is an important regulator of bone cell physiology. We studied the interaction of vitamin D metabolites on the hormonal-activated Ca message system in the osteoblastic cell line UMR-106. The acute rise in [Ca2+]i induced by different calciotropic hormones [parathyroid hormone, prostaglandin E2 (PGE2)] was dose dependently blunted by 1,25-dihydroxyvitamin D [1,25(OH)2D3; half-maximal inhibitory concn approximately 5 x 10(-11) M] and was initially observed after 8 h of preincubation. The 1,25(OH)2D3 metabolite of vitamin D was two orders of magnitude more potent than 24,25(OH)2D3 and 25(OH)D3. To discern between an effect of 1,25(OH)2D3 on hormonal-induced Ca2+ entry through the plasma membrane channel vs. release of Ca2+ from internal stores, we suspended fura-2-loaded cells in Mn2+ rather than Ca2+ buffers. In cells preincubated with 1,25(OH)2D3, [Ca2+]i release (indicated by [Ca2+]i transient) was significantly blunted, whereas Mn2+ influx (indicating Ca2+ flux across the plasma membrane) was unaltered, suggesting a selective effect of 1,25(OH)2D3 on hormonal-activated release of Ca2+ from intracellular stores. 1,25(OH)2D3 also inhibited the PGE2-induced production of inositol 1,4,5-trisphosphate. We conclude that, in osteoblasts, chronic (hours) incubation with 1,25(OH)2D3 leads to attenuated stimulation of the [Ca2+]i transduction pathway by calciotropic hormones. This effect of 1,25(OH)2D3 may provide a cellular basis for the synergism between the effects of vitamin D and calciotropic hormones at the bone level.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Parathyroid Hormone Fragment 1-34 bovine, ≥97% (HPLC), powder