Merck
A method for the rapid detection of the zopiclone degradation product 2-amino-5-chloropyridine.
Science & justice : journal of the Forensic Science Society (2000-05-05)
J H Galloway, I D Marsh, C M Newton, A R Forrest
RÉSUMÉ
Zopiclone (Zimovane) is a cyclopyrrolone compound which exhibits hypnotic and sedative effects while also exhibiting anticonvulsant and muscle relaxant activities. The detection and quantification of zopiclone is difficult. It has a high molecular weight compared to most other commonly used drugs, therapeutic levels are not high, and it is unstable in nucleophilic solvents. A degradation product of zopiclone, 2-amino-5-chloropyridine (ACP) together with a method for its detection using high-performance liquid chromatography with diode array detection has been described previously. An account is presented of a simple method for the detection of ACP using gas chromatography with mass selective detection (GC/MS) which will facilitate detection of zopiclone use as part of a routine screen.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-Amino-5-chloropyridine, 98%