Merck
Antioxidant activity of (+)-bergenin: a phytoconstituent isolated from the bark of Sacoglottis uchi Huber (Humireaceae).
Organic & biomolecular chemistry (2008-07-18)
Heitor A De Abreu, Izandina Aparecida Dos S Lago, Gilmar P Souza, Dorila Piló-Veloso, Hélio A Duarte, Antônio Flávio de C Alcântara
RÉSUMÉ
(+)-Bergenin (1) was isolated from Sacoglottis uchi, a species of vegetable found in the Amazon region and popularly used for the treatment of several hepatic problems. The structure of 1 was fully characterized using IR, GC-MS and NMR (1D and 2D) analyses. This phytoconstituent has been used as an oriental folk medicine for the treatment of many diseases and shows antihepatotoxic properties. Tests with beta-carotene, DPPH and a heterogeneous Fenton system were carried out, confirming the antioxidant activity of 1. Theoretical calculations were performed to investigate the formation of the radical derivatives of 1 using *H, *OH, *CH(3), and *CCl(3) as initiator radicals. DFT thermodynamic calculations showed that the methoxyl group (O-6-CH(3)) is the most favorable site for radical attack. Frontier molecular orbital analysis showed that nucleophilic radical attack is favored on the aromatic ring of 1 where the LUMO is localized, with antibonding character with respect to the O-6-CH(3) bond. The possibilities of attack at other sites on 1 were investigated in detail in order to understand the regiospecificity of this reaction.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Bergenin, ≥95% (LC/MS-ELSD)