Merck
One-step casting of Laemmli discontinued sodium dodecyl sulfate-polyacrylamide gel electrophoresis gel.
Analytical biochemistry (2011-11-01)
Xiaoqiang Wu, Hisashi Koiwa
RÉSUMÉ
A modified Laemmli sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) protocol is described. The new method saves 30 min for gel casting without loss of the resolution power of Laemmli gel. In this method, both the upper and lower gels can be cast at the same time because the lower gel contains 10% glycerol, which generates higher density in the lower gel than in the upper gel.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Laemmli Lysis-buffer, non smelling