Merck
  • Effects of Investigational Moisturizers on the Skin Barrier and Microbiome following Exposure to Environmental Aggressors: A Randomized Clinical Trial and Ex Vivo Analysis.
Effects of Investigational Moisturizers on the Skin Barrier and Microbiome following Exposure to Environmental Aggressors: A Randomized Clinical Trial and Ex Vivo Analysis.
Journal of clinical medicine (2023-09-28)
Dan-Qi Wang, Xi Li, Ru-Yi Zhang, Chao Yuan, Bo Yan, Philippe Humbert, Zhe-Xue Quan
RÉSUMÉ
The skin microbiota barrier participates in skin barrier function in addition to the physical, chemical, and immunological protective barriers, and is affected by environmental aggressors and skincare regimens. To better understand the exact effects of real-life environmental conditions on the skin and determine the protective methods, this study investigates the effects of three topical cosmetic moisturizers (water gel moisturizers with/without yeast extract (Moisturizers K and C) and a thick-emulsion cream moisturizer (Moisturizer L)) on clinical and skin microbiome endpoints in the presence of environmental aggressors during an 8-week, randomized controlled, triple-blind clinical trial with 110 participants, and molecular- as well as biomarker-level endpoints on ex vivo skin explants after exposure to simulate urban environmental conditions. The results show that all moisturizers are well-tolerated and improve skin barrier function and surface moisture content from the baseline, and the improvement is maintained at the last analysis point (3 days after trial completion). Compared with the untreated control areas (samples taken from the upper chest), treatment with Moisturizer K prevented a reduction in bacterial and fungal richness, and increased the change ratio of the relative abundance of commensal bacteria, such as Staphylococcus epidermidis and Ralstonia, at the treated sites (samples taken from the forehead). Moreover, Moisturizer K-treated ex vivo skin explants had higher levels of caspase 14 (a marker of skin barrier function), collagen I, and elastin (structure components), and lower levels of aryl hydrocarbon receptor (AHR; activated by air pollutants) and interleukin-6 (IL-6) than those in explants treated with other moisturizers and in the untreated areas of the skin. These results suggest that a skin postbiotic moisturizer with yeast extract supports the regulation of the skin's microbiome balance and may provide a holistic barrier (involving skin microbiome, physical, chemical, and immune barriers) to protect the skin against environmental aggressors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps monoclonal anti-collagène de type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
ANTI-AHR antibody produced in mouse, clone 3B9, purified immunoglobulin, buffered aqueous solution