- Analyzing mammalian female meiosis.
Analyzing mammalian female meiosis.
Methods in molecular biology (Clifton, N.J.) (2009-08-18)
Martha Susiarjo, Carmen Rubio, Patricia Hunt
PMID19685334
RÉSUMÉ
The goal of this chapter is twofold: First, to acquaint the reader with the problems inherent in analyzing mammalian female meiosis and, second, to provide a step-by-step approach to mastering the necessary techniques. Although the methods presented are for use in the human and mouse, with subtle alterations the same techniques should be applicable to most mammalian species.