Merck
A novel peptide from funnel web spider venom, omega-Aga-TK, selectively blocks, P-type calcium channels.
Biochemical and biophysical research communications (1993-10-15)
T Teramoto, M Kuwada, T Niidome, K Sawada, Y Nishizawa, K Katayama
RÉSUMÉ
In the course of purification of omega-Aga-IVA, a specific P-type calcium channel blocker, from the venom of Agelenopsis aperta we discovered a novel peptide. This peptide, named omega-agatoxin Tsukuba (omega-Aga-TK), also blocked P-type channels and was twelve times more abundant in the venom than omega-Aga-IVA. omega-Aga-TK was purified to homogeneity by a two-step reverse-phase HPLC procedure. Its amino acid sequence is 71% identical to that of omega-Aga-IVA. omega-Aga-TK has a negatively charged N-terminus, whereas omega-Aga-IVA has a positively charged one. Electrophysiological data indicate that omega-Aga-TK is a potent and selective inhibitor of P-type channels.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ω-Agatoxin TK, ~98% (HPLC), powder